Cannon Four-Bulb Shear Dilution Viscometer, Cannon Instrument
Supplier: Cannon
Certificates
About this item
The Cannon Four-Bulb Shear Dilution Viscometer Five Fold Dilution type measures intrinsic viscosities extrapolated to zero shear rate with negligible kinetic energy correction
Available calibrated (CUSDC) or uncalibrated (CUSDU). Minimum sample volume 20 mL. Minimum bath depth of 11".