Anti-FCER1A Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: AER-37 (CRA1)]
Catalog # 10765-836
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide
- Antigen Symbol:FCER1A
- Clonality:Monoclonal
- Clone:AER-37 (CRA1)
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Size:25 tests
- Cross Adsorption:No
- Form:liquid
- Gene ID:P12319
- Antigen Synonyms:FCER1A
- Storage Buffer:Phosphate-buffered aqueous solution, ≤0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2.
- Storage Temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Cat. No.:10765-836
- Supplier no.:77-172
Specifications
About this item
The AER-37 (CRA1) antibody binds to the FC epsilon receptor I alpha subunit (FceR1a), a 50 kDA transmembrane protein. It is part of a tetrameric receptor complex composed from one alpha subunit, one beta subunit, and two gamma subunits. The FceR1a binds and is upregulated by the presence of IgE. It plays a role in prompting the allergic reactions mediated by IgE.
Type: Primary
Antigen: FCER1A
Clonality: Monoclonal
Clone: AER-37 (CRA1)
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, kappa
Reactivity: Human