Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Csf1r Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: AFS98]
Anti-Csf1r Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: AFS98]
Catalog # 10765-790
Supplier:  Prosci
Anti-Csf1r Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: AFS98]
Catalog # 10765-790
Supplier:  Prosci
Supplier Number:  76-880

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    colony stimulating factor 1 receptor
  • Antigen Symbol:
    Csf1r
  • Clonality:
    Monoclonal
  • Clone:
    AFS98
  • Conjugation:
    PE (Phycoerythrin)
  • Flow Cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Mouse
  • Size:
    0.025 mg
  • Cross Adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    P09581
  • Antigen Synonyms:
    Csf1r
  • Storage Buffer:
    Phosphate-buffered aqueous solution, ≤0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2.
  • Storage Temperature:
    The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
    4 °C
  • Purification:
    The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
  • Cat. No.:
    10765-790
  • Supplier no.:
    76-880

Specifications

About this item

The AFS98 monoclonal antibody specifically reacts with the mouse CD115 molecule, a 150 kDa receptor for the colony stimulating factor (CSF-1) or macrophage CFS (M-CFS), expressed by some epithelial cells, monocytes, osteoclasts, and macrophages. The colony stimulating factor-1 regulates the proliferation and the differentiation of the monocytic lineage cells. The AFS98 antibody can be used to identify myeloid lineage cells.

Type: Primary
Antigen: Csf1r
Clonality: Monoclonal
Clone: AFS98
Conjugation: PE
Epitope:
Host: Rat
Isotype: Rat IgG2a, kappa
Reactivity: Mouse