Anti-IFNG Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: NIB42]
Catalog # 10765-294
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:interferon, gamma
- Antigen Symbol:IFNG
- Clonality:Monoclonal
- Clone:NIB42
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 tests
- Cross Adsorption:No
- Neutralisation:Yes
- Form:liquid
- Gene ID:P01579
- Antigen Synonyms:IFNG
- Storage Buffer:Phosphate-buffered aqueous solution, ≤0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2.
- Storage Temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Cat. No.:10765-294
- Supplier no.:77-033
Specifications
About this item
The NIB42 monoclonal antibody specifically reacts with human interferon gamma, a multifunctional cytokine that is produced mainly by activated T and NK cells. Interferon gamma has anti-viral and anti-parasitic properties and plays an important role in the activation, growth, and differentiation of macrophages, B and T lymphocytes, and natural killer cells. It interacts synergically with other cytokines, such as TNF-alpha, to inhibit proliferation of normal and transformed cells. The NIB42 antibody neutralizes IFN-gamma.
Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: NIB42
Conjugation: PE
Epitope:
Host: Mouse
Isotype: Mouse IgG1, kappa
Reactivity: Human