Anti-TLR2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: mT2.7]
Catalog # 10765-090
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:toll-like receptor 2
- Antigen Symbol:Tlr2
- Clonality:Monoclonal
- Clone:mT2.7
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Size:0.025 mg
- Cross Adsorption:No
- Form:liquid
- Gene ID:Q9QUN7
- Antigen Synonyms:Tlr2
- Storage Buffer:Phosphate-buffered aqueous solution, ≤0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2.
- Storage Temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Cat. No.:10765-090
- Supplier no.:76-951
Specifications
About this item
The mT2.7 monoclonal antibody specifically reacts with mouse CD282, the Toll-like receptor 2 (TLR2). It is a type I transmembrane signaling receptor containing IL-1 receptor like intracellular domain and leucine-rich repeats (LRR) in the extracellular domain. CD282 is expressed on monocytes, macrophages, dendritic cells, and the RAW264.7 cell line and is involved distinguishing bacterial lipoproteins.
Type: Primary
Antigen: Tlr2
Clonality: Monoclonal
Clone: mT2.7
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, kappa
Reactivity: Mouse