Anti-CCR7 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 3D12]
Catalog # 10765-048
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:chemokine (C-C motif) receptor 7
- Antigen Symbol:CCR7
- Clonality:Monoclonal
- Clone:3D12
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:25 tests
- Cross Adsorption:No
- Form:liquid
- Gene ID:P32248
- Antigen Synonyms:CCR7
- Storage Buffer:Phosphate-buffered aqueous solution, ≤0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2.
- Storage Temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Cat. No.:10765-048
- Supplier no.:76-930
Specifications
About this item
The 3D12 monoclonal antibody specifically reacts with human CD197 (CCR7 or EBI-1), a seven-transmembrane G-protein-coupled. It is the receptor for the secondary lymphoid-tissue chemokine (SLC or 6CKine), CCL19, and CCL21. CD197 is expressed on subsets of B and T cells, and mature dendritic cells. The monoclonal 3D12 antibody recognizes the N-terminus of the receptor.
Type: Primary
Antigen: CCR7
Clonality: Monoclonal
Clone: 3D12
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: Mouse IgG1, kappa
Reactivity: Human