Anti-TLR2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: TL2.1]
Catalog # 10765-082
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:toll-like receptor 2
- Antigen Symbol:TLR2
- Clonality:Monoclonal
- Clone:TL2.1
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:25 tests
- Cross Adsorption:No
- Form:liquid
- Gene ID:O60603
- Antigen Synonyms:TLR2
- Storage Buffer:Phosphate-buffered aqueous solution, ≤0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2.
- Storage Temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Cat. No.:10765-082
- Supplier no.:76-947
Specifications
About this item
The TL2.1 monoclonal antibody specifically reacts with human CD282, the Toll-like receptor 2 (TLR2). It is a type I transmembrane signaling receptor containing IL-1 receptor like intracellular domain and leucine-rich repeats (LRR) in the extracellular domain. CD282 is expressed on peripheral blood monocytes and is involved distinguishing bacterial lipoproteins. The TL2.1 antibody blocks the receptor and immunoprecipitates TLR2 from human mammary epithelial cells (HMEC) and peripheral blood mononuclear cells (PBMC).
Type: Primary
Antigen: TLR2
Clonality: Monoclonal
Clone: TL2.1
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, kappa
Reactivity: Human