To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:hemagglutinin
- Antigen Symbol:HA
- Clonality:Monoclonal
- Clone:7B9B2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Virus
- Western Blot:Yes
- Size:0.02 mg
- Environmentally Preferable:
- Cross Adsorption:No
- Form:Liquid
- Gene ID:Q692M2
- Antigen Synonyms:Hemagglutinin [7B9B2] ,
- Storage Buffer:Hemagglutinin Monoclonal Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage Temperature:Hemagglutinin monoclonal antibody can be stored at –20 °C, stable for one year.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:A peptide corresponding to 13 amino acids in the middle of the Hemagglutinin protein.
- Purification:Hemagglutinin Monoclonal Antibody is immunoaffinity chromotography purified IgG.
- Cat. No.:10752-016
- Supplier no.:PM-4009_S
Specifications
About this item
Hemagglutinin Monoclonal Antibody: Influenza A virus is a major public health threat, killing more than 30,000 people per year in the USA. Novel influenza virus strains caused by genetic drift and viral recombination emerge periodically to which humans have little or no immunity, resulting in devastating pandemics. Influenza A can exist in a variety of animals; however it is in birds that all subtypes can be found. These subtypes are classified based on the combination of the virus coat glycoproteins hemagglutinin (HA) and neuraminidase (NA) subtypes. During 1997, an H5N1 avian influenza virus was determined to be the cause of death in 6 of 18 infected patients in Hong Kong. The more recent virulent strain of H5N1 is now seen in Africa and Europe, as well as in southeast Asia. There is some evidence of human to human spread of this virus, but it is thought that the transmission efficiency was fairly low. HA interacts with cell surface proteins containing oligosaccharides with terminal sialyl residues. Virus isolated from a human infected with the H5N1 strain in 1997 could bind to oligosaccharides from human as well as avian sources, indicating its species-jumping ability. While efforts were made to use relatively conserved regions of the viral sequence as the antigen, the influenza virus genome has drifted somewhat from what was first reported. However, this antibody was able to recognize peptides derrived from viruses from Indonesian human patients infected in 2007.
Type: Primary
Antigen: hemagglutinin
Clonality: Monoclonal
Clone: 7B9B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus