Anti-SEC62 Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
SEC62 Antibody: SEC62 is an integral membrane protein located in the rough endoplasmic reticulum (ER) and is part of the SEC61-SEC62-SEC63 complex that is the central component of the protein translocation apparatus of the ER membrane. It is speculated that SEC61-SEC62-SEC63 may perform post-translational protein translocation into the ER and might also perform the backward transport of ER proteins that are subject to the ubiquitin-proteasome-dependent degradation pathway. Silencing of this gene with RNAi dramatically reduced the migration and invasive potential of numerous tumor cell lines, suggesting that it may be an attractive target for therapy of various tumors.
Type: Primary
Antigen: SEC62
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Documents
10751-414.pdf
Datasheet
Specifications
- Size:0.02 mg
- Cross Adsorption:No
- Clonality:Polyclonal
- Purification:SEC62 Antibody is affinity chromatography purified via peptide column.
- Form:Liquid
- Storage Buffer:SEC62 Antibody is supplied in PBS containing 0.02% sodium azide.
- ImmunoFluorescence:Yes
- Isotype:IgG
- Host:Rabbit
- Concentration:1 mg/mL
- Cat. No.:10751-414
- Reactivity:Human|Rat|Mouse
- Western Blot:Yes
- Shipping Temperature:4 °C
- Antibody Type:Primary
- Antigen Symbol:SEC62
- Gene ID:Q99442
- Immunogen:SEC62 antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human SEC62. The immunogen is located within amino acids 270 - 320 of SEC62.
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:SEC62
- Antigen Name:SEC62 homolog (S. cerevisiae)
- ELISA:Yes
- Storage Temperature:SEC62 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



