Anti-ZC3H12C Rabbit Polyclonal Antibody
Catalog # 10750-886
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:zinc finger CCCH-type containing 12C
- Antigen Symbol:ZC3H12C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.02 mg
- Epitope:near the N-Terminus
- Cross Adsorption:No
- Form:Liquid
- Gene ID:Q9C0D7
- Antigen Synonyms:ZC3H12C
- Storage Buffer:ZC3H12C Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage Temperature:ZC3H12C antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:ZC3H12C antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human ZC3H12C. The immunogen is located within the first 50 amino acids of ZC3H12C.
- Purification:ZC3H12C Antibody is affinity chromatography purified via peptide column.
- Cat. No.:10750-886
- Supplier no.:5677_S
Specifications
About this item
ZC3H12C Antibody: ZC3H12C, also known as MCPIP3, is a member of a family of novel CCCH-zinc finger proteins that includes ZC3H12A, a protein that is thought to be involved in macrophage activation, host immunity and inflammatory diseases. Similar to ZC3H12A, ZC3H12C expression in macrophages is highly increased after treatment with lipopolysaccharide (LPS), suggesting it also may play a role in host immunity and inflammatory response.
Type: Primary
Antigen: ZC3H12C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat