To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:nonstructural protein 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Virus
- Size:0.02 mg
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross Adsorption:No
- Form:Liquid
- Gene ID:Q2L6Z3
- Storage Buffer:Avian Influenza Nonstructural Protein 1 Antibody is supplied in PBS containing 0.02% sodium azide.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Avian Influenza Nonstructural Protein 1 antibody was raised against a synthetic peptide corresponding to 14 amino acids at the carboxy terminus of the Avian Influenza Nonstructural Protein 1 protein. Efforts were made to use relatively conserved regions of the viral sequence as the antigen. The immunogen is located within the last 50 amino acids of Avian Influenza Nonstructural Protein 1.
- Purification:Avian Influenza Nonstructural Protein 1 Antibody is affinity chromatography purified via peptide column.
- Cat. No.:10750-568
- Supplier no.:3915_S
Avian Influenza Nonstructural Protein 1 Antibody: Influenza A virus is a major public health threat, killing more than 30, 000 people per year in the USA. Novel influenza virus strains caused by genetic drift and viral recombination emerge periodically to which humans have little or no immunity, resulting in devastating pandemics. Influenza A can exist in a variety of animals; however, it is in birds that all subtypes, including the so-called "avian flu" or H5N1, can be found. These subtypes are classified based on the combination of the virus coat glycoproteins hemagglutinin (HA) and neuraminidase (NA) subtypes. One of the less studied proteins encoded by, but not incorporated in, the influenza virus is the nonstructural protein (NS) 1. NS1 counters cellular antiviral activities and acts as a virulence factor. It can bind to double-stranded RNA and sequester it from 2'-5'OAS, preventing the activation of the RNAse L, which normally acts to degrade RNA and prevent virus replication. NS1 also binds to and inhibits the anti-viral protein kinase PKR.
Type: Primary
Antigen: nonstructural protein 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus