Anti-SARS Rabbit Polyclonal Antibody
Catalog # 10749-694
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Envelope small membrane protein
- Antigen Symbol:E
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Virus
- Size:0.02 mg
- Epitope:C-Terminal
- Cross Adsorption:No
- Form:Liquid
- Gene ID:P59637
- Antigen Synonyms:E
- Storage Buffer:SARS Envelope Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage Temperature:SARS Envelope antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:SARS envelope antibody was raised against a synthetic peptide corresponding to amino acids at the carboxy terminus of the SARS envelope protein. The immunogen is located within the last 50 amino acids of SARS Envelope.
- Purification:SARS Envelope Antibody is affinity chromatography purified via peptide column.
- Cat. No.:10749-694
- Supplier no.:3533_S
Specifications
About this item
SARS Envelope Antibody: A novel coronavirus has recently been identified as the causative agent of SARS (Severe Acute Respiratory Syndrome). Coronaviruses are a major cause of upper respiratory diseases in humans. The genomes of these viruses are positive-stranded RNA approximately 27-31kb in length. SARS infection can be mediated by the binding of the viral spike protein, a glycosylated 139 kDa protein and the major surface antigen of the virus, to the angiotensin-converting enzyme 2 (ACE2) on target cells. This binding can be blocked by a soluble form of ACE2.
Type: Primary
Antigen: E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus