- Antibody Type:Primary
- Antigen Name:BCL2-like 13 (apoptosis facilitator)
- Antigen Symbol:BCL2L13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.02 mg
- Cross Adsorption:No
- Form:Liquid
- Gene ID:Q9BXK5
- Antigen Synonyms:BCL2L13
- Storage Buffer:Bcl-rambo Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage Temperature:Bcl-rambo antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Bcl-rambo antibody was raised against a 15 amino acid synthetic peptide from near the center of human Bcl-rambo. The immunogen is located within amino acids 280 - 330 of Bcl-rambo.
- Purification:Bcl-rambo Antibody is affinity chromatography purified via peptide column.
- Cat. No.:10749-498
- Supplier no.:3023_S
Bcl-rambo Antibody: Apoptosis plays a major role in normal organism development, tissue homeostasis, and removal of damaged cells. Disruption of this process has been implicated in a variety of diseases such as cancer. Members of the Bcl-2 family are known to be critical regulators of this process. These proteins are characterized by the presence of several conserved motifs termed Bcl-2 homology (BH) domains. A novel, widely expressed member termed Bcl-rambo was recently identified. This protein is localized to mitochondria in mammalian cells and its overexpression induces apoptosis which could be blocked by co-expression of inhibitor of apoptosis proteins (IAPs) such as XIAP, cIAP1, and cIAP2. Bcl-rambo shows overall homology to the anti-apoptotic members containing BH motifs, but unlike Bcl-2, the C-terminal membrane anchor of Bcl-rambo is preceded by a unique 250 amino acid insertion. This region by itself can induce apoptosis more efficiently than the Bcl-2 homology regions, suggesting that Bcl-rambo may be important other pro-apoptotic pathways.
Type: Primary
Antigen: BCL2L13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat