Specifications
- Antibody Type:Primary
- Antigen Name:tumor necrosis factor (ligand) superfamily, member 13
- Antigen Symbol:TNFSF13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.02 mg
- Cross Adsorption:No
- Form:Liquid
- Gene ID:O75888
- Antigen Synonyms:TNFSF13
- Storage Buffer:APRIL Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular Weight:27 kDa
- Storage Temperature:APRIL antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping Temperature:4 °C
- Immunogen:APRIL antibody was raised against a synthetic peptide corresponding to amino acids near the center of human APRIL. The immunogen is located within amino acids 50 - 100 of APRIL.
- Purification:APRIL Antibody is affinity chromatography purified via peptide column.
- Cat. No.:10749-348
- Supplier no.:2223_S
Specifications
About this item
APRIL Antibody: Members in the TNF superfamily regulate immune responses and induce apoptosis. A novel member in the TNF family was recently identified by several groups and designated APRIL (for a proliferation-inducing ligand), TALL-2 (for TNF- and ApoL-related Leukocyte-expressed Ligand 2), and TRDL-1alpha (for TNF related death ligand 1alpha ) in human and mouse. Two receptors for APRIL were recently identified and designated TACI and BCMA. APRIL stimulates B and T cell proliferation, triggers humoral immune responses, activates NF-kappa B, and induces cell death. APRIL and its close relative of BlyS and their receptors BCMA and TACI are involved in diseases of autoimmunity and cancer.
Type: Primary
Antigen: TNFSF13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human