Specifications
- Antibody Type:Primary
- Antigen Name:endonuclease G
- Antigen Symbol:ENDOG
- Clonality:Monoclonal
- Clone:7F2D7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.02 mg
- Cross Adsorption:No
- Form:Liquid
- Gene ID:Q14249
- Antigen Synonyms:ENDOG
- Storage Buffer:EndoG Monoclonal Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage Temperature:EndoG monoclonal antibody can be stored at –20 °C, stable for one year.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Recombinant protein corresponding to amino acids 51 - 140 of human EndoG.
- Purification:EndoG Monoclonal Antibody is immunoaffinity chromotography purified IgG.
- Cat. No.:10749-164
- Supplier no.:PM-4581_S
Specifications
About this item
EndoG Monoclonal Antibody: The fragmentation of nuclear DNA is a hallmark of apoptotic cell death. The activities of caspase and nuclease are involved in the DNA fragmentation. Caspase-activated deoxyribonuclease (CAD), also termed DNA fragmentation factor (DFF40), is one such nuclease, and is capable of inducing DNA fragmentation and chromatin condensation after cleavage by caspase-3 of its inhibitor ICAD/DFF45. Caspase and CAD independent DNA fragmentation also exists. Recent studies demonstrated that another nuclease, endonuclease G (EndoG), is specifically activated by apoptotic stimuli and is able to induce nucleosomal fragmentation of DNA independently of caspase and DFF/CAD. EndoG is a mitochondrion-specific nuclease that translocates to the nucleus and cleaves chromatin DNA during apoptosis. The homologue of mammalian EndoG is the first mitochondrial protein identified to be involved in apoptosis in C. elegans. EndoG also cleaves DNA in vitro.
Type: Primary
Antigen: ENDOG
Clonality: Monoclonal
Clone: 7F2D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat