Anti-SLC29A2 Rabbit Polyclonal Antibody
10749-080
: Prosci
- Antibody Type:Primary
- Antigen Name:solute carrier family 29 (equilibrative nucleoside transporter), member 2
- Antigen Symbol:SLC29A2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.02 mg
- Cross Adsorption:No
- Form:Liquid
- Gene ID:Q14542
- Antigen Synonyms:SLC29A2
- Storage Buffer:SLC29A2 antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular Weight:Predicted: 50 kDa Observed: 50 kDa
- Storage Temperature:SLC29A2 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:SLC29A2 antibody was raised against a 19 amino acid peptide near the center of human SLC29A2. The immunogen is located within amino acids 250 - 300 of SLC29A2.
- Purification:SLC29A2 antibody is affinity chromatography purified via peptide column.
- Cat. No.:10749-080
- Supplier no.:8127_S
SLC29A2 is a member of the equilibrative nucleoside transporter family which plays a key role in nucleoside and nucleobase uptake for salvage pathways of nucleotide synthesis. SLC29A2 is a transmembrane glycoprotein that mediates the cellular uptake of nucleosides from the surrounding medium. As a nucleoside transporter, SLC29A2 plays an important role in the uptake of nucleoside-based anti-cancer drugs; polymorphisms of point mutations in the gene encoding this protein may affect the efficacy of these drugs.
Type: Primary
Antigen: SLC29A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human