Anti-ADRM1 Rabbit Polyclonal Antibody
Catalog # 10749-074
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:adhesion regulating molecule 1
- Antigen Symbol:ADRM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.02 mg
- Cross Adsorption:No
- Form:Liquid
- Gene ID:Q16186
- Antigen Synonyms:ADRM1
- Storage Buffer:ADRM1 antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular Weight:Predicted: 45 kDa Observed: 48 kDa
- Storage Temperature:ADRM1 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:ADRM1 antibody was raised against a 19 amino acid peptide near the center of human ADRM1. The immunogen is located within amino acids 210 - 260 of ADRM1.
- Purification:ADRM1 antibody is affinity chromatography purified via peptide column.
- Cat. No.:10749-074
- Supplier no.:8119_S
Specifications
About this item
The adhesion-regulating molecule 1 (ADRM1) is thought to be involved in protein recruitment and cell adhesion and functions as a proteasomal ubiquitin receptor. ADRM1 promotes the activity of UCH-L5 and plays a role in lymphocyte homing. The carboxy-terminal domain of mammalian ADRM1 serves to bind and enhance the isopeptidase activity of UCHL5/UCH37, perhaps serving as a mechanism to accelerate ubiquitin chain disassembly. ADRM1 expression is induced by IFN-gamma in some cancer cell lines and its expression is upregulated in other metastatic cells, suggesting a role in carcinogenesis.
Type: Primary
Antigen: ADRM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat