Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-NA Mouse Monoclonal Antibody [clone: 10C5F12]
  10748-944
 :  Prosci
undefined
Anti-NA Mouse Monoclonal Antibody [clone: 10C5F12]
  10748-944
 :  Prosci
 :  PM-5919_S

 

  • Antibody Type:
    Primary
  • Antigen Name:
    hemagglutinin
  • Antigen Symbol:
    NA
  • Clonality:
    Monoclonal
  • Clone:
    10C5F12
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Virus
  • Size:
    0.02 mg
  • Cross Adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    B1AGS8
  • Antigen Synonyms:
    NA
  • Storage Buffer:
    Seasonal H1N1 Neuraminidase Monoclonal Antibody is supplied in PBS containing 0.02% sodium azide.
  • Storage Temperature:
    Seasonal H1N1 Neuraminidase monoclonal antibody can be stored at –20 °C, stable for one year.
  • Concentration:
    1 mg/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    Mouse monoclonal neuraminidase antibody was raised against a synthetic peptide from the seasonal influenza (H1N1) neuraminidase protein.
  • Purification:
    Seasonal H1N1 Neuraminidase Monoclonal Antibody is immunoaffinity chromotography purified IgG.
  • Cat. No.:
    10748-944
  • Supplier no.:
    PM-5919_S

 

 

Seasonal H1N1 Neuraminidase Monoclonal Antibody: Influenza A virus has one of sixteen possible hemagglutinin (HA) surface proteins and one of nine possible neuraminidase (NA) surface proteins. In early 2009, a novel H1N1 swine-origin influenza (S-OIV) A virus was identified in specimens obtained from patients in Mexico and the United States. The genetic make-up of this swine flu virus is unlike any other: it is an H1N1 strain that combines a triple assortment first identified in 1998 including human, swine, and avian influenza with two new pig H3N2 virus genes from Eurasia, themselves of recent human origin.

Type: Primary
Antigen: NA
Clonality: Monoclonal
Clone: 10C5F12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Virus