Specifications
- Antibody Type:Primary
- Antigen Name:corticotropin releasing hormone
- Antigen Symbol:CRH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.02 mg
- Epitope:N-Terminal
- Cross Adsorption:No
- Form:Liquid
- Gene ID:P06850
- Antigen Synonyms:CRH
- Storage Buffer:CRH antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular Weight:Predicted: 22 kDa Observed: 29 kDa
- Storage Temperature:CRH antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:CRH antibody was raised against a 16 amino acid peptide near the amino terminus of human CRH. The immunogen is located within amino acids 30 - 80 of CRH.
- Purification:CRH antibody is affinity chromatography purified via peptide column.
- Cat. No.:10748-928
- Supplier no.:7797_S
Specifications
About this item
Corticotropin-releasing hormone (CRH) is secreted by the paraventricular nucleus (PVN) of the hypothalamus in response to stress. In addition to production in the hypothalamus, this protein is also synthesized in peripheral tissues, such as T lymphocytes and is highly expressed in the placenta where it is a marker that determines the length of gestation and the timing of parturition and delivery. A rapid increase in circulating levels of the hormone occurs at the onset of parturition, suggesting that, in addition to its metabolic functions, this protein may act as a trigger for parturitio. Marked reduction in this protein has been observed in association with Alzheimer disease.
Type: Primary
Antigen: CRH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human