Order Entry
Puerto Rico
ContactUsLinkComponent
Phospholipase D (PLD) Activity Colorimetric Assay Kit, BioVision
Phospholipase D (PLD) Activity Colorimetric Assay Kit, BioVision
Catalog # 10186-986
Supplier:  Avantor
CAS Number:  
undefined
Phospholipase D (PLD) Activity Colorimetric Assay Kit, BioVision
Catalog # 10186-986
Supplier:  Avantor
CAS Number:  

Specifications

  • Size:
    100 assays
  • Species Reactivity:
    Mammalian samples
  • Shelf Life:
    12 Months
  • Sample Size:
    Animal tissues: liver, brain, kidney, etc., Cell culture: Adherent or suspension cells
  • Storage Temperature:
    -20°C
  • Cat. No.:
    10186-986
  • Supplier no.:
    K725-100

Specifications

About this item

Highly Sensitive Assay, HTS

Detection method- Absorbance (OD 570 nm)
Applications- - Measurement of PLD activity in various tissues/cells
- Analyzing intracellular signaling
- Screening of PLD inhibitors

Phospholipase D (PLD) is a key player in phospholipid metabolism. PLD hydrolyzes the phosphodiester bond of the glycerophospholipids, resulting in the production of phosphatidic acid and a free headgroup. PLD activity regulates the actin cytoskeleton, vesicle trafficking for secretion and endocytosis, and receptor signaling. Abnormalities in PLD activity and expression have been associated with Alzheimer's disease, stroke, cancer and other brain disorders. BioVision’s Phospholipase D Activity Assay Kit provides a simple and sensitive method of measuring PLD activity using colorimetry (OD 570 nm). In this assay, PLD cleaves choline from Phosphatidylcholine. The free choline is then oxidized by PLD enzyme mix to generate an intermediate which reacts with PLD Probe to generate color (OD 570 nm). This high-throughput adaptable assay kit can detect PLD activity as low as 1.0 mU/ml in a variety of samples.