Order Entry
Puerto Rico
ContactUsLinkComponent
Glucose Colorimetric Assay Kit II, BioVision
Glucose Colorimetric Assay Kit II, BioVision
Catalog # 10186-956
Supplier:  Avantor
CAS Number:  
undefined
Glucose Colorimetric Assay Kit II, BioVision
Catalog # 10186-956
Supplier:  Avantor
CAS Number:  

Specifications

  • Size:
    100 assays
  • Species Reactivity:
    Mammalian samples
  • Shelf Life:
    12 Months
  • Sample Size:
    Cell and tissue culture supernatants, urine, plasma and serum, as well as many other biological fluids, fermentation media, food samples etc.
  • Storage Temperature:
    -20°C
  • Cat. No.:
    10186-956
  • Supplier no.:
    K686-100

Specifications

About this item

For samples having reducing substances

Detection method- Absorbance (450 nm)
Species reactivity- Mammalian
Application- The kit can detect glucose concentrations in the range of 20µM – 10mM.

Glucose is an important fuel source to generate the universal energy molecule ATP. Serum glucose level is a key diagnostic parameter for many metabolic disorders. BioVision’s Glucose Assay Kit II provides direct measurement of glucose in various biological samples (e.g., serum, plasma, other body fluids, food, growth media, etc.). In this assay, glucose is specifically oxidized to generate a product which reacts with a dye to generate color (λ = 450 nm) whose intensity is proportional to glucose concentration. The method is rapid, simple, sensitive, and suitable for high throughput. This assay is particularly suitable for serum and urine samples since it is unaffected by reducing substances which can interfere with other suppliers offering oxidase-based kits. The assay is also suitable for monitoring glucose level during fermentation and glucose feeding in protein expression processes. The kit can detect glucose concentrations in the range of 20µM – 10mM.