Cannon-Ubbelohde Dilution Viscometer, Cannon Instrument
Supplier: Cannon
Certificates
About this item
The Cannon-Ubbelohde Dilution Viscometer measures intrinsic viscosity of transparent Newtonian liquids according to ASTM D445 and ISO 3104
Available calibrated (CUDC) and uncalibrated (CUDU). Minimum sample volume of 8 mL. Minimum bath depth of 12".