Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant Dr41 (from HEK293 Cells)
Human Recombinant Dr41 (from HEK293 Cells)
Catalog # 10798-474
Supplier:  Prosci
CAS Number:  
undefined
Human Recombinant Dr41 (from HEK293 Cells)
Catalog # 10798-474
Supplier:  Prosci
Supplier Number:  96-745
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    0.1 mg
  • Tag sequence:
    His Tag
  • Protein Synonyms:
    TRAIL-R1|DR4|TNFRSF10A|APO2|CD261
  • Protein/Peptide Name:
    Dr41
  • Purity:
    >92% as determined by SDS-PAGE.
  • Molecular Weight:
    24 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested Applications:
    Western Blot
  • Cat. No.:
    10798-474
  • Supplier no.:
    96-745

Specifications

About this item

Tumor necrosis factor receptor superfamily member 10A (TNFRSF10A) is also known as TNF-related apoptosis-inducing ligand receptor 1 (TRAIL-R1), Death receptor 4 (DR4), CD261 and APO2, which belongs to TNF superfamily. TRAILR1 / TNFRSF10A contains 1 death domain and 3 TNFR-Cys repeats. TNFRSF10A / DR4 is widely expressed and high levels are found in spleen, peripheral blood leukocytes, small intestine and thymus, but also in K-562 erythroleukemia cells, MCF-7 breast carcinoma cells and activated T-cells. APO2 / TNFRSF10A is receptor for the cytotoxic ligand TNFSF10 / TRAIL. The adapter molecule FADD recruits caspase-8 to the activated receptor.

The resulting death-inducing signaling complex (DISC) performs caspase-8 proteolytic activation which initiates the subsequent cascade of caspases (aspartate-specific cysteine proteases) mediating apoptosis. TRAILR-1 / DR4 / CD261 promotes the activation of NF-kappa-B.