Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Human Recombinant CD951 (from HEK293 Cells)

Human Recombinant CD951 (from HEK293 Cells)

0.05 mg
Catalog # 10798-178
Supplier:   Prosci
Supplier #:  96-301
Human Recombinant CD951 (from HEK293 Cells)
Catalog # 10798-178
Supplier:  Prosci
CAS Number:  
Human Recombinant CD951 (from HEK293 Cells)
Human Recombinant CD951 (from HEK293 Cells)

0.05 mg

CAS Number:  
Price
$732.35
Log in for availability
Each (50µG)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The Fas is also known as FAS receptor (FasR), apoptosis antigen 1 (APO-1 or APT), cluster of differentiation 95 (CD95) or tumor necrosis factor receptor superfamily member 6 (TNFRSF6). is a death receptor on the surface of cells that leads to programmed cell death (apoptosis). It is one of two apoptosis pathways, the other being the mitochondrial pathway. FasR is located on chromosome 10 in humans and 19 in mice. Similar sequences related by evolution (orthologs) are found in most mammals. Fas forms the death-inducing signaling complex (DISC) upon ligand binding. Membrane-anchored Fas ligand trimer on the surface of an adjacent cell causes trimerization of Fas receptor. This event is also mimicked by binding of an agonistic Fas antibody, though some evidence suggests that the apoptotic signal induced by the antibody is unreliable in the study of Fas signaling. To this end, several clever ways of trimerizing the antibody for in vitro research have been employed.Upon ensuing death domain (DD) aggregation, the receptor complex is internalized via the cellular endosomal machinery. This allows the adaptor molecule FADD to bind the death domain of Fas through its own death domain. Recently, Fas has also been shown to promote tumor growth, since during tumor progression, it is frequently downregulated or cells are rendered apoptosis resistant. Cancer cells in general, regardless of their Fas apoptosis sensitivity, depend on constitutive activity of Fas. This is stimulated by cancer-produced Fas ligand for optimal growth.

Documents

Specifications

Specifications

  • Size:
    0.05 mg
  • Cat. No.:
    10798-178
  • Molecular Weight:
    18.2 kDa
  • Protein/Peptide Type:
    Recombinant
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Purity:
    >92% as determined by SDS-PAGE.
  • Tested Applications:
    Western Blot
  • Protein Synonyms:
    FasR|FAS|FASTM|APO1|ALPS1A|TNFRSF6|CD95|APT1|FAS1
  • Protein/Peptide Name:
    CD951
  • Tag sequence:
    His Tag
  • Species:
    Human
  • Source:
    HEK293 cells

Specifications