BAX® System PCR Assay for Enterobacter sakazakii (Cronobacter spp.), Hygiena™, Qualicon Diagnostics LLC
Catalog # 10754-908
Supplier: Hygiena
CAS Number:
Specifications
- Sample Type:Powdered Infant Formula, Dry Dairy, Soy Ingredients, Food Production Environmentals
- Processing Time:3.5 hours
- Cat. No.:10754-908
- Response time:20-22 hours depending upon food type, sample volume, and enrichment media
- Target group:Cronobacter spp.
- Supplier no.:D11801836
- No. of tests:96
Specifications
About this item
The Hygiena™ BAX® System PCR Assay for Enterobacter sakazakii (Cronobacter spp.) produces clear yes-or-no results after a fast processing time in the BAX® System Q7 instrument.
Delivering next-day results, the DuPont BAX System PCR Assay for Enterobacter sakazakii (Cronobacter spp.) produces clear yes-or-no results in 3.5 hours of processing in the BAX System Q7 instrument. Developed in partnership with the Nestle Research Centre, this assay offers automated DNA analysis instead of subjective plate counts.
The Hygiena BAX System is an automated detection system that uses the power of the polymerase chain reaction (PCR) to detect harmful bacteria and other microbes in food ingredients, finished products and environmental surfaces.
Ordering information: Includes PCR tubes with tablets, optical caps, protease, and lysis buffer. This assay can only be used with the BAX System Q7 instrument.