Order Entry
Puerto Rico
ContactUsLinkComponent
BAX® System Real-Time PCR Assay for Shigella, Hygiena™, Qualicon Diagnostics LLC
BAX® System Real-Time PCR Assay for Shigella, Hygiena™, Qualicon Diagnostics LLC
Catalog # 10754-886
Supplier:  Hygiena
CAS Number:  
undefined
BAX® System Real-Time PCR Assay for Shigella, Hygiena™, Qualicon Diagnostics LLC
Catalog # 10754-886
Supplier:  Hygiena
Supplier Number:  KIT2007
CAS Number:  

Specifications

  • Sample Type:
    Frankfurters, Raw Pork, Dessert Gelatin, Ready-To-Eat Meals
  • Processing Time:
    65 minutes
  • Cat. No.:
    10754-886
  • Response time:
    8-20 hours, depending on food type, sample volume and enrichment media
  • Target group:
    Shigella spp.
  • Supplier no.:
    D14812018
  • No. of tests:
    96

Specifications

About this item

The Hygiena™ BAX® System PCR Assay for Shigella produces clear yes-or-no results after a fast processing time in the BAX® System Q7 instrument.

The Hygiena BAX System Real-Time PCR Assay for Shigella is a screening assay designed to quickly eliminate negative results, reducing the number of samples that must be confirmed. This assay produces clear yes-or-no results after about 65 minutes of processing in the BAX System Q7 instrument. The BAX System Real-Time PCR Assay for Shigella offers fewer presumptive positive results than traditional methods, which means reduced hands-on time and cost for culture confirmations.

The Hygiena BAX System is an automated detection system that uses the power of the polymerase chain reaction (PCR) to detect harmful bacteria and other microbes in food ingredients, finished products and environmental surfaces.

Ordering information: Includes PCR tubes with tablets, optical caps, protease, and lysis buffer. This assay can only be used with the BAX System Q7 instrument.