Order Entry
Puerto Rico
ContactUsLinkComponent
 
BAX® System Real-Time PCR Assay for Staphylococcus aureus, Hygiena™, Qualicon Diagnostics LLC
Catalog # 10754-890
Supplier:  Hygiena
CAS Number:  
undefined
BAX® System Real-Time PCR Assay for Staphylococcus aureus, Hygiena™, Qualicon Diagnostics LLC
Catalog # 10754-890
Supplier:  Hygiena
Supplier Number:  KIT2020
CAS Number:  

Specifications

  • Sample Type:
    Powdered Infant Formula, Ground Beef, Soy Protein Isolate
  • Processing Time:
    70 minutes
  • Cat. No.:
    10754-890
  • Response time:
    22-48 hours depending upon food type, sample volume, and enrichment media
  • Target group:
    Staphylococcus aureus
  • Supplier no.:
    D12762689
  • No. of tests:
    96

Specifications

About this item

The Hygiena™ BAX® System PCR Assay for Staphylococcus aureusproduces clear yes-or-no results after a fast processing time in the BAX® System Q7 instrument.

The HygienaBAX System Real-Time PCR Assay for Staphylococcus aureus produces clear, yes-or-no, next-day results after 70 minutes of processing in the BAX System Q7 instrument. Certified as an AOAC-RI Performance Tested MethodSM for powdered infant formula, ground beef and soy protein isolate.

The Hygiena BAX System is an automated detection system that uses the power of the polymerase chain reaction (PCR) to detect harmful bacteria and other microbes in food ingredients, finished products and environmental surfaces.

Ordering information: Includes PCR tubes with tablets, optical caps, protease, and lysis buffer. This assay can only be used with the BAX System Q7 instrument.