ContactUsLinkComponent
BAX® System PCR Assay for Escherichia coli O157:H7, Hygiena™, Qualicon Diagnostics LLC
BAX® System PCR Assay for Escherichia coli O157:H7, Hygiena™, Qualicon Diagnostics LLC
Supplier:  Hygiena
Certificates
undefined
BAX® System PCR Assay for Escherichia coli O157:H7, Hygiena™, Qualicon Diagnostics LLC
Supplier:  Hygiena
Specifications
  • Sample Type:
    Raw Ground Beef, Beef Trim, Produce
  • Processing Time:
    65 minutes