Anti-BAG5 Goat Polyclonal Antibody
89303-662
: Genetex
- Antibody Type:Primary
- Antigen Name:BCL2 Associated Athanogene 5
- Antigen Symbol:BAG5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Bovine,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:9529
- Antigen Synonyms:BAG family molecular chaperone regulator 5| BCL2 associated athanogene 5|BAG5_HUMAN.|BAG 5|KIAA0873|BAG-5
- Storage Buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage Temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5mg/ml
- Immunogen:DMGNQHPSISRLQ-C
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:89303-662
- Supplier no.:GTX89733
Goat polyclonal antibody to BAG5 (N Terminus)
Preliminary experiments gave a strong band at 35kDa with fainter bands at 48kDa and 70kDa in Human Heart lysates at 1ug/ml. Please note that currently we cannot find an explanation in the literature for the band we observe given the predicted size of approx. 66kDa according to NP_004864. We would appreciate any feedback from people in the field - have any results been reported with other antibodies/lysates? Have any further splice variants/modified forms been reported? Variants (NP_004864.1; NP_001015048.1) encode the same isoform.
Type: Primary
Antigen: BAG5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Bovine, Dog, Rat