- Antibody Type:Primary
- Antigen Name:Perilipin 4
- Antigen Symbol:S3-12 / KIAA1881 (perilipin 4)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μg
- Gene ID:729359
- Antigen Synonyms:KIAA1881|S3-12
- Storage Buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage Temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5mg/ml
- Immunogen:C-RTTDNGGEQTALSPQE
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:89303-594
- Supplier no.:GTX89103
Goat polyclonal antibody to S3-12 / KIAA1881
Preliminary experiments gave bands at approx 45kDa and 50kDa in Human Adipose lysates after 0.1µg/ml antibody staining. Please note that currently we cannot find an explanation in the literature for the bands we observe given the calculated size of 134kDa according to NP_001073869.1. Both detected bands were successfully blocked by incubation with the immunizing peptide (and BLAST results with the immunizing peptide sequence did not identify any other proteins to explain the additional bands). We would appreciate any feedback from people in the field - have any results been reported with other antibodies/lysates? Have any further splice variants/modified forms been reported?
Type: Primary
Antigen: S3-12 / KIAA1881 (perilipin 4)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human