Anti-PITX3 Goat Polyclonal Antibody
Catalog # 89296-654
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Paired-like HomeoDomain 3
- Antigen Symbol:PITX3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:5309
- Antigen Synonyms:paired-like homeodomain 3|homeobox protein PITX3|PTX3|MGC12766
- Storage Buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage Temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5mg/ml
- Immunogen:PEHGCKGQEHSD
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:89296-654
- Supplier no.:GTX88644
Specifications
About this item
Goat polyclonal antibody to PITX3
Approx 40kDa band observed in Human Brain (Cerebellum and Cerebral Cortex) lysates (calculated MW of 31.8kDa according to NP_005020.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Coulon et al, J Biol Chem. 2007 Nov 9;282(45):33192-200.; PMID: 17848564). Recommended concentration: 0.3-1µg/ml. This antibody is not excpected to cross-react with PITX1 and PITX2.
Type: Primary
Antigen: PITX3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human