Anti-APOBPEC3C Goat Polyclonal Antibody
Catalog # 89296-602
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Apolipoprotein B mRNA Editing Enzyme Catalytic Polypeptide-Like 3C
- Antigen Symbol:APOBPEC3C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:27350
- Antigen Synonyms:ARDC2|ARDC4|APOBEC1L|ARP5|A3C|bK150C2.3|PBI
- Storage Buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage Temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5mg/ml
- Immunogen:C-RLLKRRLRESLQ
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:89296-602
- Supplier no.:GTX88604
Specifications
About this item
Goat polyclonal antibody to APOBEC3C (C-term)
Approx. 22kDa band observed in lysates of cell line DAUDI (calculated MW of 22.8kDa according to NP_055323.2). Recommended concentration: 0.5-1.5µg/ml.
Type: Primary
Antigen: APOBPEC3C
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human