Anti-BST2 Goat Polyclonal Antibody
Catalog # 89296-100
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Tetherin / CD317 (bone marrow stromal cell Antigen 2)
- Antigen Symbol:BST2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:684
- Antigen Synonyms:CD317|TETHERIN
- Storage Buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage Temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5mg/ml
- Immunogen:C-ELTEAQKGFQD
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:89296-100
- Supplier no.:GTX88171
Specifications
About this item
Goat polyclonal antibody to Tetherin / CD317
Approx 30kDa band observed in Human Spleen and Testis lysates (calculated MW of 19.8kDa according to NP_004326.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Rollason, J Cell Sci. 2007 Nov 1;120(Pt 21):3850-8.; PMID: 17940069). Recommended concentration: 1-3µg/ml.
Type: Primary
Antigen: BST2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human