Anti-IMP1 (insulin-like growth Factor 2 mRNA binding Protein 1) Goat Polyclonal Antibody
Catalog # 89295-966
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Insulin-like Growth Factor 2 mRNA binding Protein 1
- Antigen Symbol:IGF2BP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Bovine,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:10642
- Antigen Synonyms:IMP-1|CRDBP|VICKZ1|ZBP1|CRD-BP|IMP1
- Storage Buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage Temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5mg/ml
- Immunogen:C-EKVFAEHKISYSGQ
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:89295-966
- Supplier no.:GTX88062
Specifications
About this item
Goat polyclonal antibody to IMP1
Approx 65kDa band observed in lysates of cell line K562 (calculated MW of 63.5kDa according to NP_006537.3;). Recommended concentration: 0.3-1µg/ml. This antibody is expected to recognize both reported isoforms (NP_006537.3; NP_001153895.1).
Type: Primary
Antigen: IMP1 (insulin-like growth Factor 2 mRNA binding Protein 1)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Bovine, Dog, Rat