Anti-MAT1 Rabbit Polyclonal Antibody
89291-468
: Genetex
- Antibody Type:Primary
- Antigen Name:Mnat Cdk-Activating Kinase Assembly Factor 1
- Antigen Symbol:MAT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:4331
- Antigen Synonyms:CAP35|RNF66|MAT1|TFB3
- Storage Buffer:Phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Storage Temperature:Store at -20 degrees Celcius. Product is stable for at least 1 year.
- Immunogen:The antiserum was produced against synthesized peptide derived from human MAT1.
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen.
- Cat. No.:89291-468
- Supplier no.:GTX87969
Rabbit polyclonal antibody to MAT1
We recommend the following starting dilutions: WB: 1:500~1:1000 ELISA: 1:20000Optimal dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: MAT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat