Anti-SCN1A Rabbit Polyclonal Antibody
Catalog # 89291-392
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Sodium Channel Voltage-gated Type I Alpha
- Antigen Symbol:SCN1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:6323
- Antigen Synonyms:SMEI|SCN1|FEB3|HBSCI|FEB3A|NAC1|GEFSP2|EIEE6|FHM3|Nav1.1
- Storage Buffer:Phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Storage Temperature:Store at -20 degrees Celcius. Product is stable for at least 1 year.
- Immunogen:The antiserum was produced against synthesized peptide derived from human Sodium Channel.
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen.
- Cat. No.:89291-392
- Supplier no.:GTX87930
Specifications
About this item
Rabbit polyclonal antibody to Sodium Channel-pan
We recommend the following starting dilutions: WB: 1:500~1:1000 IHC: 1:50~1:100 ELISA: 1:20000Optimal dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: SCN1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat