Anti-DDB1 Rabbit Polyclonal Antibody
Catalog # 89300-140
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:damage-specific DNA binding protein 1, 127kDa
- Antigen Symbol:DDB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:25 μl
- Gene ID:1642
- Antigen Synonyms:XAP1|UV-DDB1|XPE-BF|XPE|XPCE|DDBA
- Storage Buffer:0.01% sodium azide.
- Storage Temperature:+4 °C or –20 °C, Avoid repeated freezing and thawing.
- Concentration:85mg/ml
- Immunogen:Synthetic peptide - KLH conjugated
- Purification:This antibody was purified using Delipidation and Defibrination
- Cat. No.:89300-140
- Supplier no.:GTX85639
Specifications
About this item
Rabbit polyclonal antibody to DDB1
WB (1:500 - 1:1000), IP, ELISA (1:2000 - 1:10000), IHC-P (1:500)
Type: Primary
Antigen: DDB1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human