Anti-GSTT2 Mouse Monoclonal Antibody [clone: 8A10]
Catalog # 89273-588
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Glutathione S-transferase Theta 2
- Antigen Symbol:GSTT2
- Clonality:Monoclonal
- Clone:8A10
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Gel Shift:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:2953
- Storage Buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0.02% sodium azide.
- Storage Temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human GSTT2 (NP_000845) produced in HEK293T cell.
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Cat. No.:89273-588
- Supplier no.:GTX84398
Specifications
About this item
Mouse monoclonal antibody [8A10] to GSTT2
Recommended Starting Dilutions: WB 1:500~2000 ICC/IF 1:100 Flow Cytometry 1:100Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: GSTT2
Clonality: Monoclonal
Clone: 8A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human