Anti-H6PD Mouse Monoclonal Antibody [clone: 2A7]
Catalog # 89273-582
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Hexose-6-phosphate dehydrogenase
- Antigen Symbol:H6PD
- Clonality:Monoclonal
- Clone:2A7
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Gel Shift:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Monkey,Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:9563
- Antigen Synonyms:CORTRD1|GDH|Glucose 1-dehydrogenase|G6PDH
- Storage Buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0.02% sodium azide.
- Storage Temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human H6PD (NP_004276) produced in HEK293T cell.
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Cat. No.:89273-582
- Supplier no.:GTX84395
Specifications
About this item
Mouse monoclonal antibody [2A7] to H6PD
Recommended Starting Dilutions: WB 1:1000 IHC 1:50 ICC/IF 1:100 Flow Cytometry 1:100Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: H6PD
Clonality: Monoclonal
Clone: 2A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey