To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Interleukin 1 Family Member 6
- Antigen Symbol:IL1-F6
- Clonality:Monoclonal
- Clone:1D11
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Gel Shift:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:27179
- Antigen Synonyms:REGalpha|FIL1E|PA28A|FIL1|IL1F6|IFI5111|IL1(EPSILON)|FIL1(EPSILON)|PA28alpha|IL-1F6
- Storage Buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0.02% sodium azide.
- Storage Temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human IL1F6 (NP_055255) produced in HEK293T cell.
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Cat. No.:89273-392
- Supplier no.:GTX84299
Mouse monoclonal antibody [1D11] to IL1F6
Recommended Starting Dilutions: WB 1:2000 ICC/IF 1:100,Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: IL1F6
Clonality: Monoclonal
Clone: 1D11
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human