Anti-PARP16 Mouse Monoclonal Antibody [clone: 2C8]
Catalog # 89272-684
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Poly (ADP-ribose) polymerase 16
- Antigen Symbol:PARP16
- Clonality:Monoclonal
- Clone:2C8
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Gel Shift:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 μl
- Gene ID:54956
- Antigen Synonyms:C15orf30|ARTD15|pART15
- Storage Buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0.02% sodium azide.
- Storage Temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Protein expressed in 293T cell transfected with human PARP16 expression vector .
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Cat. No.:89272-684
- Supplier no.:GTX83943
Specifications
About this item
Mouse monoclonal antibody [2C8] to PARP16
Recommended Starting Dilutions:Flow 1:500Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: PARP16
Clonality: Monoclonal
Clone: 2C8
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human