Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-PDE4A Mouse Monoclonal Antibody [clone: 7B11]
Anti-PDE4A Mouse Monoclonal Antibody [clone: 7B11]
  89272-644
 :  Genetex
Anti-PDE4A Mouse Monoclonal Antibody [clone: 7B11]
  89272-644
 :  Genetex
 :  GTX83922
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Phosphodiesterase 4A, cAMP-specific
  • Antigen Symbol:
    PDE4A
  • Clonality:
    Monoclonal
  • Clone:
    7B11
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Gel Shift:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    5141
  • Antigen Synonyms:
    PDE46|DPDE2|PDE4|ANAPC6|CUT9|APC6
  • Storage Buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0.02% sodium azide.
  • Storage Temperature:
    For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
  • Immunogen:
    Full length human recombinant protein of human PDE4A (NP_006193) produced in HEK293T cell.
  • Purification:
    Purified from mouse ascites fluids by Protein A/G affinity chromatography
  • Cat. No.:
    89272-644
  • Supplier no.:
    GTX83922

 

 

Mouse monoclonal antibody [7B11] to PDE4A

Recommended Starting Dilutions: WB 1:2000 IHC 1:50 ICC/IF 1:100 Flow Cytometry 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PDE4A
Clonality: Monoclonal
Clone: 7B11
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human