Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-PEX5 Mouse Monoclonal Antibody [clone: 6E9]
Catalog # 89272-610
Supplier:  Genetex
Anti-PEX5 Mouse Monoclonal Antibody [clone: 6E9]
Catalog # 89272-610
Supplier:  Genetex
Supplier Number:  GTX83905
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Peroxisomal Biogenesis Factor 5
  • Antigen Symbol:
    PEX5
  • Clonality:
    Monoclonal
  • Clone:
    6E9
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Gel Shift:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Monkey,
    Human,
    Dog,
    Rat
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    5830
  • Antigen Synonyms:
    PBD2A|PTS1-BP|PTS1R|PBD2B|PXR1
  • Storage Buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0.02% sodium azide.
  • Storage Temperature:
    For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
  • Immunogen:
    Full length human recombinant protein of human PEX5 (NP_000310) produced in HEK293T cell.
  • Purification:
    Purified from mouse ascites fluids by Protein A/G affinity chromatography
  • Cat. No.:
    89272-610
  • Supplier no.:
    GTX83905

Specifications

About this item

Mouse monoclonal antibody [6E9] to PEX5

Recommended Starting Dilutions: WB 1:500~2000 ICC/IF 1:100 Flow Cytometry 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PEX5
Clonality: Monoclonal
Clone: 6E9
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Dog, Monkey, Rat