Anti-PYCR2 Mouse Monoclonal Antibody [clone: 2F11]
Catalog # 89272-310
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Pyrroline-5-carboxylate Reductase 2
- Antigen Symbol:PYCR2
- Clonality:Monoclonal
- Clone:2F11
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:29920
- Antigen Synonyms:HLD10|P5CR2
- Storage Buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0.02% sodium azide.
- Storage Temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human PYCR2 (NP_037460) produced in HEK293T cell.
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Cat. No.:89272-310
- Supplier no.:GTX83754
Specifications
About this item
Mouse monoclonal antibody [2F11] to PYCR2
Recommended Starting Dilutions: WB 1:2000 ICC/IF 1:100 Flow Cytometry 1:100Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: PYCR2
Clonality: Monoclonal
Clone: 2F11
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human