Anti-PYCR2 Mouse Monoclonal Antibody [clone: 2A5]
89272-308
: Genetex
- Antibody Type:Primary
- Antigen Name:Pyrroline-5-carboxylate Reductase 2
- Antigen Symbol:PYCR2
- Clonality:Monoclonal
- Clone:2A5
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Gel Shift:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Monkey,Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:29920
- Antigen Synonyms:HLD10|P5CR2
- Storage Buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0.02% sodium azide.
- Storage Temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human PYCR2 (NP_037460) produced in HEK293T cell.
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Cat. No.:89272-308
- Supplier no.:GTX83753
Mouse monoclonal antibody [2A5] to PYCR2
Recommended Starting Dilutions: WB 1:500~2000 ICC/IF 1:100,Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: PYCR2
Clonality: Monoclonal
Clone: 2A5
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Monkey