Anti-SLC7A8 Mouse Monoclonal Antibody [clone: 4H10]
Catalog # 89272-046
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Solute Carrier Family 7 Member 8
- Antigen Symbol:SLC7A8
- Clonality:Monoclonal
- Clone:4H10
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Gel Shift:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 μl
- Gene ID:23428
- Antigen Synonyms:HBO1|ZC2HC7|HBOA|MYST2
- Storage Buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0.02% sodium azide.
- Storage Temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human SLC7A8 (NP_036376) produced in HEK293T cell.
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Cat. No.:89272-046
- Supplier no.:GTX83621
Specifications
About this item
Mouse monoclonal antibody [4H10] to SLC7A8
Recommended Starting Dilutions:WB 1:500 IHC 1:50 Flow 1:500Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: SLC7A8
Clonality: Monoclonal
Clone: 4H10
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human