Anti-KSHV ORF62 Mouse Monoclonal Antibody [clone: 5B7B6]
Catalog # 89303-404
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Symbol:KSHV ORF62
- Clonality:Monoclonal
- Clone:5B7B6
- ELISA:Yes
- Host:Mouse
- Reactivity:Virus
- Size:100 µL
- Cat. No.:89303-404
- Supplier no.:GTX83235
Specifications
About this item
Type: Primary
Antigen: KSHV ORF62
Clonality: Monoclonal
Clone: 5B7B6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus