Anti-K5 Mouse Monoclonal Antibody [clone: 7H8C4]
Catalog # 89287-414
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Keratin 5, Type Ii
- Antigen Symbol:K5
- Clonality:Monoclonal
- Clone:7H8C4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:3852
- Antigen Synonyms:DDD|KRT5|CK5|EBS2|KRT5A
- Storage Buffer:Ascitic fluid containing 0.03% sodium azide.
- Storage Temperature:Store at 4℃, for long term storage, store at -20℃.
- Immunogen:The immunogen corresponding to CK5 expressed in E. Coli.
- Cat. No.:89287-414
- Supplier no.:GTX83079
Specifications
About this item
Mouse monoclonal antibody [7H8C4] to Cytokeratin (Pan)
Recommended Starting Dilutions:ELISA: 1/10000.WB: 1/500 - 1/2000.IHC: 1/200 - 1/1000.ICC: 1/200 - 1/1000.Not yet tested in other applications.Determining optimal working dilutions by titration test.
Type: Primary
Antigen: K5
Clonality: Monoclonal
Clone: 7H8C4
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human