To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:NMDAR1 Splice Variant C2
- Antigen Symbol:NDMAR1 SV C2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:24408
- Storage Temperature:For long-term storage, aliquot and store at -20 C. Avoid repeated freeze/thaw cycles.
- Immunogen:Synthetic peptide corresponding to amino acid residues specific to the NR1 subunit, C2 splice variant insert conjugated to KLH
- Purification:Antigen affinity purification
- Cat. No.:89284-058
- Supplier no.:GTX82639
Specifications
About this item
Rabbit polyclonal antibody to NMDA NR1 Subunit, Splice Variant C2
Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: NDMAR1 SV C2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat