Anti-POSTN Rabbit Polyclonal Antibody
Catalog # 89284-022
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Periostin
- Antigen Symbol:POSTN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:50706
- Antigen Synonyms:PLF|OSF-2|A630052E07Rik|Osf2|PN|AI747096
- Storage Buffer:100 ul in PBS
- Storage Temperature:For long-term storage, aliquot and store at -20 C. Avoid repeated freeze/thaw cycles.
- Immunogen:Synthetic peptide corresponding to amino acid residues from the fasciclin domain 1 of mouse periostin
- Purification:Antigen affinity purification
- Cat. No.:89284-022
- Supplier no.:GTX82615
Specifications
About this item
Rabbit polyclonal antibody to Periostin (pan)
Type: Primary
Antigen: POSTN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: 0